2-Fluor-4-pyridinboronsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Fluor-4-pyridinylboronsäure, 2-Fluorpyridin-4-boronsäure
H27630.06
H27630.06EA
481
EUR
H27630.06
2-Fluor-4-pyridinboronsäure 95%
2-Fluoro-4-pyridineboronic acid
Purity: 95%
CAS No.: 401815-98-3
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Storage Temperature: -30°C to -10°C
Melting Point: ca 195° dec.
MDL No.: MFCD04112534
Formel:
C₅H₅BFNO₂ Molecular Weight: 140,91 g/mol Schmelzpunkt: ∼ 195 °C (Zers.) |
MDL:
MFCD04112534 CAS-Nummer: 401815-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...