α-(N,N-Dimethylamino)phenylacetonitrile 97%

Lieferant: Thermo Fisher Scientific
Gefahr

H27631.14 H27631.06
H27631.14EA 105 EUR
H27631.14 H27631.06
α-(N,N-Dimethylamino)phenylacetonitrile 97%
alpha-(N,N-Dimethylamino)phenylacetonitrile

Purity: 97% CAS No.: 827-36-1 Molecular Formula: C10H12N2 Formula Weight: 160.22 Flash Point: 104°(219°F) Refractive Index: 1.514 Boiling Point: 88-90°/2mm UN No.: UN3276 MDL No.: MFCD00013810

Formel: C₁₀H₁₂N₂
Siedepunkt: 88…90 °C (2 mmHg)
Dichte: 0,997
Flammpunkt: 104 °C (219 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013810
CAS-Nummer: 827-36-1
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR