4-Brom-2,6-dinitrotoluol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
H27634.06
H27634.06EA
167
EUR
H27634.06
4-Brom-2,6-dinitrotoluol ≥97%
4-Brom-2,6-dinitrotoluol
Purity: 97%
CAS No.: 95192-64-6
Molecular Formula: C7H5BrN2O4
Formula Weight: 261.03
Melting Point: 86-89°
UN No.: 2811
MDL No.: MFCD07357280
Formel:
C₇H₅BrN₂O₄ Molecular Weight: 261,03 g/mol Schmelzpunkt: 86…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07357280 CAS-Nummer: 95192-64-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...