4-Brom-2-fluorpyridin 95%

Lieferant: Thermo Scientific
Gefahr

H27657.06
H27657.06EA 176 EUR
H27657.06
4-Brom-2-fluorpyridin 95%
4-Bromo-2-fluoropyridine

Purity: 95% CAS No.: 128071-98-7 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Refractive Index: 1.5300

Formel: C₅H₃BrFN
Molecular Weight: 175,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112504
CAS-Nummer: 128071-98-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR