Methylamin 2 M in THF

Lieferant: Thermo Scientific
Gefahr

Synonyme: Aminomethan

H27664.AE
H27664.AEEA 100 EUR
H27664.AE
Methylamin 2 M in THF
Methylamin
Formel: CH₅N
Molecular Weight: 31,06 g/mol
Dichte: 0,861
Flammpunkt: <–34 °C (–29 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008104
CAS-Nummer: 74-89-5
EINECS: 200-820-0
UN: 2924
ADR: 3,II
Merck Index: 14,6014

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR