4-Amino-5-brom-2-chlorpyridin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Brom-2-chlor-4-pyridylamin

H27674.03
H27674.03EA 147 EUR
H27674.03
4-Amino-5-brom-2-chlorpyridin 95%
4-Amino-5-bromo-2-chloropyridine

Purity: 95% CAS No.: 857730-21-3 Molecular Formula: C5H4BrClN2 Formula Weight: 207.46 Melting Point: 127-129° MDL No.: MFCD09037459

Formel: C₅H₄BrClN₂
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09037459
CAS-Nummer: 857730-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR