3-Brom-2-ethoxypyridin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H27690.03
H27690.03EA
100
EUR
H27690.03
3-Brom-2-ethoxypyridin 95%
3-Bromo-2-ethoxypyridine
Purity: 95%
CAS No.: 57883-25-7
Molecular Formula: C7H8BrNO
Formula Weight: 202.05
MDL No.: MFCD00234309
Formel:
C₇H₈BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234309 CAS-Nummer: 57883-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...