(S)-(+)-3-Aminopiperidin Dihydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-(+)-3-Piperidinamine dihydrochloride, (S)-Piperidin-3-amine dihydrochloride

H27700.03
H27700.03EA 108 EUR
H27700.03
(S)-(+)-3-Aminopiperidin Dihydrochlorid 98%
(S)-(+)-3-Aminopiperidin Dihydrochlorid

Purity: 98% CAS No.: 334618-07-4 Molecular Formula: C5H12N2.2HCl Formula Weight: 173.08 Melting Point: ca 210° dec. Optical Rotation: +1.6° (c=1 in water)

Formel: C₅H₁₄Cl₂N₂
Molecular Weight: 173,08 g/mol
Schmelzpunkt: ∼ 210 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD03427036
CAS-Nummer: 334618-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR