Diethyl allyl(3-phenyl-2-propynyl)malonate 96%
Lieferant: Thermo Scientific
|
Achtung
|
H27702.03
H27702.03EA
67.1
EUR
H27702.03
Diethyl allyl(3-phenyl-2-propynyl)malonate 96%
Diethyl allyl(3-phenyl-2-propynyl)malonate
Purity: 96%
CAS No.: 143633-91-4
Molecular Formula: C19H22O4
Formula Weight: 314.38
Refractive Index: 1.5140
MDL No.: MFCD09953493
Formel:
C₁₉H₂₂O₄ Molecular Weight: 314,38 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953493 CAS-Nummer: 143633-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...