4-Benzyloxy-5-bromo-2-chloropyrimidine 97%
Lieferant: Thermo Scientific
Synonyme:
4-(Benzyloxy)-5-bromo-2-chloropyrimidine
H27706.03
H27706.03EA
128
EUR
H27706.03
4-Benzyloxy-5-bromo-2-chloropyrimidine 97%
4-Benzyloxy-5-bromo-2-chloropyrimidine
Purity: 97%
CAS No.: 205672-19-1
Molecular Formula: C11H8BrClN2O
Formula Weight: 299.56
Melting Point: 114-116°
MDL No.: MFCD06798237
Formel:
C₁₁H₈BrClN₂O Molecular Weight: 299,56 g/mol Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798237 CAS-Nummer: 205672-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...