4,5-Dimethylthiophene-2-carboxaldehyde 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27712.03
H27712.03EA
75.6
EUR
H27712.03
4,5-Dimethylthiophene-2-carboxaldehyde 97%
4,5-Dimethylthiophene-2-carboxaldehyde
Purity: 97%
CAS No.: 5928-48-3
Molecular Formula: C7H8OS
Formula Weight: 140.20
Flash Point:
Formel:
C₇H₈OS Siedepunkt: 82…85 °C Dichte: 1,179 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093897 CAS-Nummer: 5928-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...