N-Acetyl-DL-serine ≥98%

Lieferant: Thermo Scientific

Synonyme: Ac-DL-Ser-OH

H27717.06
H27717.06EA 416 EUR
H27717.06
N-Acetyl-DL-serine ≥98%
N-Acetyl-DL-serine

Purity: 98+% CAS No.: 97-14-3 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Storage Temperature: -30°C to -10°C MDL No.: MFCD00037248

Formel: C₅H₉NO₄
Molecular Weight: 147,1 g/mol
MDL: MFCD00037248
CAS-Nummer: 97-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR