(E)-1,2-Diazendiylbis(1-piperidinylmethanon) 97%

Lieferant: Thermo Fisher Scientific
Achtung

H27719.14 H27719.06
H27719.14EA 325 EUR
H27719.14 H27719.06
(E)-1,2-Diazendiylbis(1-piperidinylmethanon) 97%
1,1'-(Azodicarbonyl)dipiperidine

Purity: 97% CAS No.: 10465-81-3 Molecular Formula: C12H20N4O2 Formula Weight: 252.32 Odour: Odourless Melting Point: 133-136° MDL No.: MFCD00010111

Formel: C₁₂H₂₀N₄O₂
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010111
CAS-Nummer: 10465-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR