1-Butyl-3-methylimidazoliumoctylsulfat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

H27720.06 H27720.18
H27720.06EA 36.5 EUR
H27720.06 H27720.18
1-Butyl-3-methylimidazoliumoctylsulfat ≥99%
1-Butyl-3-methylimidazolium octylsulphate

Purity: 99% CAS No.: 445473-58-5 Molecular Formula: C16H32N2O4S Formula Weight: 348.50 MDL No.: MFCD03788914

Formel: C₁₆H₃₂N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD03788914
CAS-Nummer: 445473-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR