3,6-Dibromo-2-chloropyridine 95%
Lieferant: Thermo Scientific
|
Gefahr
|
H27738.03
H27738.03EA
146
EUR
H27738.03
3,6-Dibromo-2-chloropyridine 95%
3,6-Dibromo-2-chloropyridine
Purity: 95%
CAS No.: 942206-18-0
Molecular Formula: C5H2Br2ClN
Formula Weight: 271.34
Melting Point: 84-86°
MDL No.: MFCD09037464
Formel:
C₅H₂Br₂ClN Molecular Weight: 271,34 g/mol Schmelzpunkt: 84…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037464 CAS-Nummer: 942206-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...