2-Fluor-5-hydroxypyridin 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Fluor-5-pyridinol
H27741.03
H27741.03EA
272
EUR
H27741.03
2-Fluor-5-hydroxypyridin 95%
2-Fluor-5-hydroxypyridin
Purity: 95%
CAS No.: 55758-32-2
Molecular Formula: C5H4FNO
Formula Weight: 113.09
Melting Point: 150-153°
MDL No.: MFCD01075040
Formel:
C₅H₄FNO Molecular Weight: 113,09 g/mol Schmelzpunkt: 150…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075040 CAS-Nummer: 55758-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...