1-Fluoro-3,5-dimethyl-2-nitrobenzene 98%

Lieferant: Thermo Fisher Scientific
Gefahr

H27762.06
H27762.06EA 247 EUR
H27762.06
1-Fluoro-3,5-dimethyl-2-nitrobenzene 98%
1-Fluoro-3,5-dimethyl-2-nitrobenzene

Purity: 98% CAS No.: 315-13-9 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Melting Point: 54-57° MDL No.: MFCD00052367

Formel: C₈H₈FNO₂
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052367
CAS-Nummer: 315-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR