N-(α)-Boc-D-tryptophanol 98%
Lieferant: Thermo Scientific
Synonyme:
Boc-D-Tryptophanol, Boc-D-Trp-OL
H27763.03
H27763.03EA
240
EUR
H27763.03
N-(α)-Boc-D-tryptophanol 98%
N-(alpha)-Boc-D-tryptophanol
Purity: 98%
CAS No.: 158932-00-4
Molecular Formula: C16H22N2O3
Formula Weight: 290.36
Melting Point: 119-121°
Optical Rotation: +53° (c=2 in acetic acid)
MDL No.: MFCD00270221
Formel:
C₁₆H₂₂N₂O₃ Molecular Weight: 290,3 g/mol Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270221 CAS-Nummer: 158932-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...