Bis(ethylcyclopentadienyl)mangan(II) ≥98%

Lieferant: Thermo Scientific

Synonyme: Bis(ethylcyclopentadienyl)manganese

H27769.06
H27769.06EA 388 EUR
H27769.06
Bis(ethylcyclopentadienyl)mangan(II) ≥98%
Bis(ethylcyclopentadienyl)mangan(II)
Formel: C₁₄H₁₈Mn
Molecular Weight: 241,236 g/mol
MDL: MFCD01862452
CAS-Nummer: 101923-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR