4,4-Difluoro-L-prolinamide hydrochloride 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27770.03
H27770.03EA
158
EUR
H27770.03
4,4-Difluoro-L-prolinamide hydrochloride 98%
4,4-Difluoro-L-prolinamide hydrochloride
Purity: 98%
CAS No.: 426844-51-1
Molecular Formula: C5H8F2N2O.HCl
Formula Weight: 186.59
Melting Point: 221-223° dec.
Optical Rotation: -31° (c=1 in water)
MDL No.: MFCD06797014
Formel:
C₅H₈F₂N₂O·HCl Schmelzpunkt: 221…223 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797014 CAS-Nummer: 426844-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...