Methyl-4-brom-2-methoxybenzoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Brom-2-methoxybenzoesäuremethylester
H27773.09
H27773.09EA
132
EUR
H27773.09
Methyl-4-brom-2-methoxybenzoat ≥98%
Methyl-4-brom-2-methoxybenzoat
Purity: 98%
CAS No.: 139102-34-4
Molecular Formula: C9H9BrO3
Formula Weight: 245.08
Flash Point:
Formel:
C₉H₉BrO₃ Molecular Weight: 245,07 g/mol Siedepunkt: 100…108 °C (0,1 mmHg) Schmelzpunkt: 32…34 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797867 CAS-Nummer: 139102-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...