2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%

Lieferant: Thermo Scientific

Synonyme: Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 2,2-Dimethyltetrahydropyran-4-one

H27774.03
H27774.03EA 124 EUR
H27774.03
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one

Purity: 95% CAS No.: 1194-16-7 Molecular Formula: C7H12O2 Formula Weight: 128.17 Storage Temperature: 2°C to 8°C Boiling Point: 60-64°/8mm MDL No.: MFCD01549337

Formel: C₇H₁₂O₂
Molecular Weight: 128,17 g/mol
Siedepunkt: 60…64 °C (8 mmHg)
MDL: MFCD01549337
CAS-Nummer: 1194-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR