N-Boc-4-oxo-L-proline methyl ester 97%
Lieferant: Thermo Scientific
Synonyme:
(S)-1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-oxo-Pro-OMe
H27782.06
H27782.06EA
251
EUR
H27782.06
N-Boc-4-oxo-L-proline methyl ester 97%
N-Boc-4-oxo-L-proline methyl ester
Purity: 97%
CAS No.: 102195-80-2
Molecular Formula: C11H17NO5
Formula Weight: 243.26
Storage Temperature: -30°C to -10°C
Melting Point: 35-40°
MDL No.: MFCD01861778
Formel:
C₁₁H₁₇NO₅ Molecular Weight: 243,259 g/mol Schmelzpunkt: 35…40 °C |
MDL:
MFCD01861778 CAS-Nummer: 102195-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...