5-Brom-2-fluorpyrimidin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27795.03
H27795.03EA
227
EUR
H27795.03
5-Brom-2-fluorpyrimidin 95%
5-Bromo-2-fluoropyrimidine
Purity: 95%
CAS No.: 62802-38-4
Molecular Formula: C4H2BrFN2
Formula Weight: 176.98
Melting Point: 90-93°
MDL No.: MFCD07787364
Formel:
C₄H₂BrFN₂ Molecular Weight: 176.98 g/mol Schmelzpunkt: 90…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787364 CAS-Nummer: 62802-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...