1,3-Diisopropylimidazoliumchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3-Di-i-propylimidazolium chloride, 1,3-Diisopropyl-1H-imidazol-3-ium chloride

H27798.03 H27798.06
H27798.03EA 63 EUR
H27798.03 H27798.06
1,3-Diisopropylimidazoliumchlorid ≥97%
1,3-Diisopropylimidazoliumchlorid

Purity: 97+% CAS No.: 139143-09-2 Molecular Formula: C9H17ClN2 Formula Weight: 188.69 Melting Point: 182-186° MDL No.: MFCD03840577

Formel: C₉H₁₇ClN₂
Molecular Weight: 188.70 g/mol
Schmelzpunkt: 182…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840577
CAS-Nummer: 139143-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR