2-Amino-3-brom-5-chlorpyridin 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin
H27799.MD
H27799.MDEA
0
EUR
H27799.MD
2-Amino-3-brom-5-chlorpyridin 95%
2-Amino-3-brom-5-chlorpyridin
Purity: 95%
CAS No.: 26163-03-1
Molecular Formula: C5H4BrClN2
Formula Weight: 207.46
Melting Point: 83-85°
MDL No.: MFCD00955627
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955627 CAS-Nummer: 26163-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...