1-Hexylboronic acid pinacol ester 98%

Lieferant: Thermo Fisher Scientific
Achtung

H27831.03 H27831.06 H27831.14
H27831.03EA 41.9 EUR
H27831.03 H27831.06 H27831.14
1-Hexylboronic acid pinacol ester 98%
1-Hexylboronic acid pinacol ester

Purity: 98% CAS No.: 86308-26-1 Molecular Formula: C12H25BO2 Formula Weight: 212.14 Flash Point: 84°(183°F) Refractive Index: 1.426 MDL No.: MFCD08457665

Formel: C₁₂H₂₅BO₂
Dichte: 1,167
Flammpunkt: 84 °C (183 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD08457665
CAS-Nummer: 86308-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR