2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde
H27844.06
H27844.06EA
104
EUR
H27844.06
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd 96%
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
Purity: 96%
CAS No.: 5604-46-6
Molecular Formula: C5H3Cl2N3O
Formula Weight: 192.00
Melting Point: ca 220°
MDL No.: MFCD03001242
Formel:
C₅H₃Cl₂N₃O Molecular Weight: 192,00 g/mol Schmelzpunkt: ∼ 220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001242 CAS-Nummer: 5604-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...