2-Fluor-3-pyridinboronsäurepinakolester 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Fluoropyridine-3-boronic acid pinacol ester, 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H27845.03
H27845.03EA 108 EUR
H27845.03
2-Fluor-3-pyridinboronsäurepinakolester 95%
2-Fluor-3-pyridinboronsäurepinakolester

Purity: 95% CAS No.: 452972-14-4 Molecular Formula: C11H15BFNO2 Formula Weight: 223.06 Melting Point: 42-45° MDL No.: MFCD06798245

Formel: C₁₁H₁₅BFNO₂
Molecular Weight: 223,05 g/mol
Schmelzpunkt: 42…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798245
CAS-Nummer: 452972-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR