2-(Methylthio)-5-pyridinboronsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27873.MD
H27873.MDEA
78.9
EUR
H27873.MD
2-(Methylthio)-5-pyridinboronsäure 95%
2-(Methylthio)-5-pyridinboronsäure
Purity: 95%
CAS No.: 321438-86-2
Molecular Formula: C6H8BNO2S
Formula Weight: 169.01
MDL No.: MFCD03788238
Formel:
C₆H₈BNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788238 CAS-Nummer: 321438-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...