5-Brom-2,3'-bipyridyl 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Bromo-2,3'-bipyridine

H27880.03
H27880.03EA 272 EUR
H27880.03
5-Brom-2,3'-bipyridyl 95%
5-Brom-2,3'-bipyridyl

Purity: 95% CAS No.: 774-53-8 Molecular Formula: C10H7BrN2 Formula Weight: 235.08 Melting Point: 75-78° MDL No.: MFCD09037420

Formel: C₁₀H₇BrN₂
Molecular Weight: 235,08 g/mol
Schmelzpunkt: 75…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09037420
CAS-Nummer: 774-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR