4-Brom-2-methylpyridin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H27884.MD H27884.03
H27884.MDEA 34.1 EUR
H27884.MD H27884.03
4-Brom-2-methylpyridin ≥97%
4-Brom-2-methylpyridin

Purity: 97% CAS No.: 22282-99-1 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 79°(174°F) Refractive Index: 1.556 Odour: Characteristic Boiling Point: 76°/14mm MDL No.: MFCD03788196

Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
Siedepunkt: 76 °C (14 mmHg)
Dichte: 1.45
Flammpunkt: 79 °C (174 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03788196
CAS-Nummer: 22282-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR