6-Bromnicotinsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Brom-3-pyridincarbonsäure, 6-Brompyridin-3-carbonsäure
H27926.14
H27926.06
H27926.14EA
346
EUR
H27926.14
H27926.06
6-Bromnicotinsäure ≥97%
6-Bromnicotinsäure
Purity: 97%
CAS No.: 6311-35-9
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Melting Point: 179-188°
MDL No.: MFCD01927100
Formel:
C₆H₄BrNO₂ Schmelzpunkt: 179…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01927100 CAS-Nummer: 6311-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...