6-Bromnicotinsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Brom-3-pyridincarbonsäure, 6-Brompyridin-3-carbonsäure

H27926.14 H27926.06
H27926.14EA 346 EUR
H27926.14 H27926.06
6-Bromnicotinsäure ≥97%
6-Bromnicotinsäure

Purity: 97% CAS No.: 6311-35-9 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: 179-188° MDL No.: MFCD01927100

Formel: C₆H₄BrNO₂
Schmelzpunkt: 179…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01927100
CAS-Nummer: 6311-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR