3,3-Dimethylcyclohexanon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H28000.06
H28000.06EA
116
EUR
H28000.06
3,3-Dimethylcyclohexanon ≥97%
3,3-Dimethylcyclohexanone
Purity: 97%
CAS No.: 2979-19-3
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 59°(138°F)
Refractive Index: 1.4470
Boiling Point: 175°
UN No.: 1224
MDL No.: MFCD00040178
Formel:
C₈H₁₄O Molecular Weight: 126,2 g/mol Siedepunkt: 175 °C Schmelzpunkt: 6 °C Dichte: 0,91 Flammpunkt: 59 °C (138 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040178 CAS-Nummer: 2979-19-3 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...