Ethyl-7-bromoheptanoate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
7-Bromoheptanoic Acid Ethyl Ester, Ethyl 7-bromoheptanoate
H28001.AA
H28001.AAEA
45.3
EUR
H28001.AA
Ethyl-7-bromoheptanoate 97%
Ethyl-7-bromoheptanoate
Purity: 97%
CAS No.: 29823-18-5
Molecular Formula: Br(CH2)6CO2CH2CH3
Formula Weight: 237.14
Flash Point:
Formel:
C₉H₁₇BrO₂ Molecular Weight: 237.14 g/mol Siedepunkt: 112 °C (5 mmHg) Dichte: 1.217 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037103 CAS-Nummer: 29823-18-5 EINECS: 249-883-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...