(4-Isopropylphenoxy)acetyl chloride 98%

Lieferant: Thermo Scientific
Gefahr

H28036.06
H28036.06EA 91.5 EUR
H28036.06
(4-Isopropylphenoxy)acetyl chloride 98%
(4-Isopropylphenoxy)acetyl chloride

Purity: 98% CAS No.: 223128-33-4 Molecular Formula: C11H13ClO2 Formula Weight: 212.67 Refractive Index: 1.5195 UN No.: UN3265 MDL No.: MFCD02628421

Formel: C₁₁H₁₃ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02628421
CAS-Nummer: 223128-33-4
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR