1,4-Bis(methoxydimethylsilyl)benzene 97%

Lieferant: Thermo Fisher Scientific
Achtung

H28165.06 H28165.03
H28165.06EA 125 EUR
H28165.06 H28165.03
1,4-Bis(methoxydimethylsilyl)benzene 97%
1,4-Bis(methoxydimethylsilyl)benzene

Purity: 97% CAS No.: 131182-68-8 Molecular Formula: C12H22O2Si2 Formula Weight: 254.47 MDL No.: MFCD09266162

Formel: C₁₂H₂₂O₂Si₂
Molecular Weight: 254.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09266162
CAS-Nummer: 131182-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR