3-Hydroxypyrazine-2-carboxamide 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: T-1105

H28250.06
H28250.06EA 132 EUR
H28250.06
3-Hydroxypyrazine-2-carboxamide 98%
3-Hydroxypyrazine-2-carboxamide

Purity: 98% CAS No.: 55321-99-8 Molecular Formula: C5H5N3O2 Formula Weight: 139.12 Melting Point: ca 270° dec. MDL No.: MFCD00233977

Formel: C₅H₅N₃O₂
Molecular Weight: 139,114 g/mol
Schmelzpunkt: ∼ 270 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00233977
CAS-Nummer: 55321-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR