5-Phenyl-o-toluidin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Methyl-[1,1'-biphenyl]-3-amine, 3-Amino-4-methylbiphenyl
H28265.03
H28265.03EA
71
EUR
H28265.03
5-Phenyl-o-toluidin 98%
5-Phenyl-o-toluidin
Purity: 98%
CAS No.: 80938-67-6
Molecular Formula: C13H13N
Formula Weight: 183.26
MDL No.: MFCD00270126
Formel:
C₁₃H₁₃N Molecular Weight: 183,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270126 CAS-Nummer: 80938-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...