4-(Boc-amino)piperidin 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

H28517.14 H28517.06 H28517.03
H28517.14EA 504 EUR
H28517.14 H28517.06 H28517.03
4-(Boc-amino)piperidin 96%
4-(Boc-amino)piperidin

Purity: 96% CAS No.: 73874-95-0 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Melting Point: 162-166° MDL No.: MFCD00798171

Formel: C₁₀H₂₀N₂O₂
Schmelzpunkt: 162…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00798171
CAS-Nummer: 73874-95-0
EINECS: 616-026-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR