4-(Boc-amino)piperidin 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
H28517.14
H28517.06
H28517.03
H28517.14EA
504
EUR
H28517.14
H28517.06
H28517.03
4-(Boc-amino)piperidin 96%
4-(Boc-amino)piperidin
Purity: 96%
CAS No.: 73874-95-0
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Melting Point: 162-166°
MDL No.: MFCD00798171
Formel:
C₁₀H₂₀N₂O₂ Schmelzpunkt: 162…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798171 CAS-Nummer: 73874-95-0 EINECS: 616-026-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...