2-Chlor-5-fluorphenol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H28567.06
H28567.06EA
49.6
EUR
H28567.06
2-Chlor-5-fluorphenol ≥98%
2-Chlor-5-fluorphenol
Purity: 98%
CAS No.: 3827-49-4
Molecular Formula: C6H4ClFO
Formula Weight: 146.55
Refractive Index: 1.5250
Odour: Phenol-like
Boiling Point: 183-185°
UN No.: 3265
MDL No.: MFCD00042524
Formel:
C₆H₄ClFO Molecular Weight: 146,55 g/mol Siedepunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042524 CAS-Nummer: 3827-49-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...