N-Methyl-1-naphthylmethylamine hydrochloride 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H28719.03
H28719.09
H28719.03EA
45.1
EUR
H28719.03
H28719.09
N-Methyl-1-naphthylmethylamine hydrochloride 98%
N-Methyl-1-naphthylmethylamine hydrochloride
Purity: 98%
CAS No.: 65473-13-4
Molecular Formula: C12H13N.HCl
Formula Weight: 207.70
Melting Point: 190-193°
MDL No.: MFCD00012555
Formel:
C₁₂H₁₃N Schmelzpunkt: 190…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012555 CAS-Nummer: 65473-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...