Kalium(4-iodphenyl)trifluoroborat 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H28758.06
H28758.03
H28758.06EA
224
EUR
H28758.06
H28758.03
Kalium(4-iodphenyl)trifluoroborat 96%
Kalium(4-iodphenyl)trifluoroborat
Purity: 96%
CAS No.: 912350-00-6
Molecular Formula: C6H4BF3IK
Formula Weight: 309.90
Melting Point: 297-299°
MDL No.: MFCD09800738
Formel:
C₆H₄BF₃IK Schmelzpunkt: 297…299 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800738 CAS-Nummer: 912350-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...