2-Fluor-4-(trifluormethyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid

H28788.06 H28788.03
H28788.06EA 331 EUR
H28788.06 H28788.03
2-Fluor-4-(trifluormethyl)phenylboronsäure ≥97%
2-Fluor-4-(trifluormethyl)phenylboronsäure

Purity: 97% CAS No.: 503309-11-3 Molecular Formula: C7H5BF4O2 Formula Weight: 207.92 Melting Point: 113-115° MDL No.: MFCD03094995

Formel: C₇H₅BF₄O₂
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094995
CAS-Nummer: 503309-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR