4-Methyl-2-(methylthio)pyrimidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H28795.06
H28795.06EA
40.7
EUR
H28795.06
4-Methyl-2-(methylthio)pyrimidin ≥98%
4-Methyl-2-(methylthio)pyrimidine
Purity: 98%
CAS No.: 14001-63-9
Molecular Formula: C6H8N2S
Formula Weight: 140.21
Flash Point: 104°(219°F)
Refractive Index: 1.5710
MDL No.: MFCD00023242
Formel:
C₆H₈N₂S Dichte: 1,01 Flammpunkt: 104 °C (219 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023242 CAS-Nummer: 14001-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...