3-Methoxybenzolsulfonamid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H28844.03
H28844.03EA
88.1
EUR
H28844.03
3-Methoxybenzolsulfonamid ≥97%
3-Methoxybenzolsulfonamid
Purity: 97%
CAS No.: 58734-57-9
Molecular Formula: C7H9NO3S
Formula Weight: 187.22
Melting Point: 131-133°
MDL No.: MFCD08704579
Formel:
C₇H₉NO₃S Molecular Weight: 187,22 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704579 CAS-Nummer: 58734-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...