4-Chlor-2-fluorphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Chlor-2-fluorbenzolboronsäure, (4-Chlor-2-fluorphenyl)metaborsäure, Dihydroxy(4-chlor-2-fluorphenyl)boran
H28872.09
H28872.09EA
182
EUR
H28872.09
4-Chlor-2-fluorphenylboronsäure ≥97%
4-Chlor-2-fluorphenylboronsäure
Purity: 97%
CAS No.: 160591-91-3
Molecular Formula: C6H5BClFO2
Formula Weight: 174.37
Melting Point: 246-250°
MDL No.: MFCD02684293
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Schmelzpunkt: 246…250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684293 CAS-Nummer: 160591-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...