4-(Methoxycarbonyl)phenylboronsäurepinakolester ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
H28942.03
H28942.06
H28942.03EA
26.6
EUR
H28942.03
H28942.06
4-(Methoxycarbonyl)phenylboronsäurepinakolester ≥97%
4-(Methoxycarbonyl)phenylboronsäurepinakolester
Purity: 97%
CAS No.: 171364-80-0
Molecular Formula: C14H19BO4
Formula Weight: 262.11
Melting Point: 77-81°
MDL No.: MFCD02179438
Formel:
C₁₄H₁₉BO₄ Schmelzpunkt: 77…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179438 CAS-Nummer: 171364-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...